Anti-SARS-CoV-2 Spike protein Mouse Monoclonal Antibody [clone: Q8]
Supplier: Genscript
Certificates
About this item
SARS-CoV-2 (Severe acute respiratory syndrome coronavirus 2), also known as 2019-nCoV, is a positive-sense single-stranded RNA virus. It caused coronavirus disease 2019 (COVID-19). SARS-CoV-2 contains glycosylated spike (S) protein, which is composed of S1 subunit and S2 subunit. The S1 contains a receptor-binding domain (RBD) that can bind to ACE2 receptor on target cells. Lineage B.1.617 has the double mutations E484Q and L452R in the spike proteins.
- High specificity
- High affinity
- High stability
Genscript offers site-specific neutralizing antibodies, neutralizing antibody standards, and control antibodies.
Type:
Antigen: SARS-CoV-2 Spike protein
Clonality: Monoclonal
Clone: Q8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Specifications
- Catalog No:
- 77007-630
- 77007-638
- Antigen Name:
- SARS-CoV-2 Spike protein
- SARS-CoV-2 Spike protein
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- Q8
- Q8
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1 kappa
- IgG1 kappa
- ELISA:
- Yes
- Yes
- Immunogen:
- SARS-CoV-2 Spike RBD protein (L452R, E484Q)
- SARS-CoV-2 Spike RBD protein (L452R, E484Q)
- Form:
- Liquid
- Liquid
- Concentration:
- 1 mg/ml
- 1 mg/ml
- Cross Adsorption:
- No
- No
- Storage Buffer:
- PBS, pH 7.2, 0.02% sodium azide
- PBS, pH 7.2, 0.02% sodium azide
- Storage Temperature:
- Store at −20 °C. This product is stable for 1 year upon receipt, when handled and stored as instructed. Avoid repeated freezing and thawing cycles
- Store at −20 °C. This product is stable for 1 year upon receipt, when handled and stored as instructed. Avoid repeated freezing and thawing cycles