About this item
SARS-CoV-2 (Severe acute respiratory syndrome coronavirus 2), also known as 2019-nCoV, is a positive-sense single-stranded RNA virus. It caused coronavirus disease in 2019 (COVID-19). SARS-CoV-2 contains glycosylated spike (S) protein, which is composed of S1 subunit and S2 subunit. The S1 contains a receptor-binding domain (RBD) that can bind to ACE2 receptor on target cells. Lineage B.1.351 (a.k.a. 20H/501Y.V2) is a variant of SARS-CoV-2. It has multiple mutations in the RBD, including K417N, E484K, N501Y. Lineage B.1.1.7 (a.k.a. 20I/501Y.V1) is also a variant of SARS-CoV-2 and has a mutation called N501Y in the RBD.SARS-CoV-2 Spike (N501) neutralizing antibody (COV2110) is produced from cell culture in vitro under conditions free from animal-derived components. The product is a humanized neutralizing antibody, recognizing N501 site of wild-type RBD. When N501 is mutated to N501Y, the mutant RBD, such as RBD (B.1.1.7) and RBD (B.1.351) as seen in Figure 1, can't be recognized by the product.
- High specificity
- High affinity
- High stability
Genscript offers site-specific neutralizing antibodies, neutralizing antibody standards, and control antibodies.
Type:
Antigen: SARS-CoV-2 RBD
Clonality: Monoclonal
Clone: COV2110
Conjugation: Unconjugated
Epitope:
Host: Human
Isotype: IgG1
Reactivity:
Specifications
- Catalog No:
- 77007-570
- 77007-508
- Antigen Name:
- SARS-CoV-2 RBD
- SARS-CoV-2 RBD
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- COV2110
- COV2110
- Host:
- Human
- Human
- Isotype:
- IgG1
- IgG1
- ELISA:
- Yes
- Yes
- Neutralisation:
- Yes
- Yes
- Immunogen:
- Recombinant SARS-CoV-2 RBD Protein
- Recombinant SARS-CoV-2 RBD Protein
- Form:
- Liquid
- Liquid
- Concentration:
- 1 mg/ml
- 1 mg/ml
- Cross Adsorption:
- No
- No
- Storage Buffer:
- Supplied in 0.2 μm filtered PBS, pH 7.2
- Supplied in 0.2 μm filtered PBS, pH 7.2
- Storage Temperature:
- Store at −20 °C. This product is stable for 1 year upon receipt, when handled and stored as instructed. Avoid repeated freezing and thawing cycles
- Store at −20 °C. This product is stable for 1 year upon receipt, when handled and stored as instructed. Avoid repeated freezing and thawing cycles