Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-Panitumumab Rabbit Polyclonal Antibody
Anti-Panitumumab Rabbit Polyclonal Antibody
Catalog # 77007-614
Supplier:  Genscript
Anti-Panitumumab Rabbit Polyclonal Antibody
Catalog # 77007-614
Supplier:  Genscript
Supplier Number:  A02063-40

Specifications

  • Antigen Name:
    Panitumumab
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Size:
    40 μg
  • Cross Adsorption:
    No
  • Form:
    Lyophilized
  • Storage Buffer:
    This product is lyophilized with PBS, pH 7.2, containing 0.02% sodium azide
  • Storage Temperature:
    The lyophilized product remains stable up to 1 year at −20 °C from date of receipt. Upon reconstitution, it can be stored for 2 - 3 weeks at 2 - 8 °C or for up to 12 months at −20 °C or below. Avoid repeated freezing and thawing cycles
  • Concentration:
    Reconstitute the lyophilized powder with deionized water (or equivalent) to a final concentration of 0.5 mg/ml
  • Immunogen:
    Panitumumab
  • Cat. No.:
    77007-614
  • Packaging:
    Vial

Specifications

About this item

Panitumumab with trade name Vectibix, is an FDA-approved drug for the treatment of patients with EGFR-expressing metastatic colorectal carcinoma with non-mutated (wild-type) KRAS after failure of fluoropyrimidine-, oxaliplatin- and irinotecan-containing chemotherapy regimens. It is a human monoclonal antibody. Panitumumab binds to the human EGF receptor (EGFR), preventing binding of activating ligands, such as the EGF and transforming growth factor alpha. GenScript anti-panitumumab antibody, pAb, Rabbit is produced from sera of a rabbit immunized with Panitumumab.

  • High specificity
  • High affinity
  • High stability

GenScript has developed a broad range of off-the-shelf anti-idiotype antibodies against blockbuster drugs to help accelerate your research of biosimilars.

Type:
Antigen: Panitumumab
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: