- Antibody Type:Primary
- Antigen Name:Protein A
- Clonality:Monoclonal
- Clone:5G10A7
- Conjugation:Biotin
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Western Blot:Yes
- Size:40 μg
- Cross Adsorption:No
- Form:Lyophilized
- Storage Buffer:Lyophilized with PBS, pH 7.4, containing 10 mg/ml BSA and 0.02% sodium azide
- Storage Temperature:It remains stable in lyophilized form for at least two years if stored at −20 °C or below. The reconstituted antibody can be stored for 2 - 3 weeks at 2 - 8 °C or for up to 12 months at −20 °C or below. Avoid repeated freezing and thawing cycles
- Concentration:Reconstitute the lyophilized powder with deionized water (or equivalent) to an final concentration of 0.5 mg/mL.
- Immunogen:Purified recombinant Protein A
- Cat. No.:77007-572
- Packaging:Vial
Protein A, a bacterial cell wall protein isolated from Staphylococcus aureus, binds to mammalian IgGs mainly through Fc regions. The protein A is most commonly bound to a chromatography resin, used for production of antibody drugs or reagents. Protein A antibody can be used for quantification of protein A in sample foods or for evaluation of protein A resin. This protein A mAb is advantageous due to its IgG1 isotype, for mouse IgG1 has weak binding capacity to protein A.GenScript protein A Antibody [biotin], mAb, mouse is protein A antibody conjugated with biotin. The conjugated antibody is suitable for detecting the expression level of protein A.
- High specificity
- High affinity
- High stability
GenScript offers highly specific monoclonal and polyclonal primary antibodies. These primary antibodies are validated for multiple applications, including Sandwich ELISA and western blot.
Type: Primary
Antigen: Protein A
Clonality: Monoclonal
Clone: 5G10A7
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: