Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-Insulin Mouse Monoclonal Antibody [clone: 6E9F1]
Anti-Insulin Mouse Monoclonal Antibody [clone: 6E9F1]
Catalog # 77007-576
Supplier:  Genscript
Anti-Insulin Mouse Monoclonal Antibody [clone: 6E9F1]
Catalog # 77007-576
Supplier:  Genscript
Supplier Number:  A01715-40
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Insulin
  • Clonality:
    Monoclonal
  • Clone:
    6E9F1
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2a kappa
  • Reactivity:
    Human
  • Size:
    40 μg
  • Cross Adsorption:
    No
  • Form:
    Lyophilized
  • Storage Buffer:
    Lyophilized with PBS, pH 7.4, containing 0.02% sodium azide. Products of 1 mg and 5 mg size are provided in liquid form
  • Storage Temperature:
    The antibody is stable in lyophilized form if stored at −20 °C or below. The reconstituted antibody can be stored for 2 - 3 weeks at 2 - 8 °C. For long term storage, aliquot and store at −20 °C or below. Avoid repeated freezing and thawing cycles
  • Concentration:
    Reconstitute the lyophilized powder with deionized water (or equivalent) to an final concentration of 0.5 mg/mL.
  • Immunogen:
    Human recombinant Insulin expressed in yeast
  • Cat. No.:
    77007-576
  • Packaging:
    Vial

Specifications

About this item

Insulin is one of the major regulatory hormones of intermediate metabolism throughout the body. It regulates the cellular uptake, utilization, and storage of glucose, amino acids, and fatty acids and inhibits the breakdown of glycogen, protein, and fat. Proinsulin is the prohormone precursor to insulin made in pancreas. It is processed by a series of proteases to form mature insulin. Mature insulin has 35 fewer amino acids; 4 are removed altogether, and the remaining 31 form the C-Peptide. The C-Peptide is abstracted from the center of the proinsulin sequence; the two other ends (alpha and β chains) remain connected by disulfide bonds. Deficiency of insulin results in diabetes mellitus, one of the leading causes of morbidity and mortality in the general population. Insulin is also present in tumors of B cell origin such as insulinoma. GenScript Insulin Antibody (6E9F1), mAb, Mouse is produced from the hybridoma resulting from fusion of SP2/0-Ag14 myeloma and B-lymphocytes obtained from mouse immunized with human recombinant Insulin expressed in yeast.

  • High specificity
  • High affinity
  • High stability

GenScript offers highly specific monoclonal and polyclonal primary antibodies. These primary antibodies are validated for multiple applications, including Sandwich ELISA and western blot.

Type: Primary
Antigen: Insulin
Clonality: Monoclonal
Clone: 6E9F1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human