Order Entry
Puerto Rico
ContactUsLinkComponent
5,5'-Divinyl-2,2'-bipyridine 97% stabilized
Irritant
5,5'-Divinyl-2,2'-bipyridine 97% stabilized
Catalog # 77035-384
Supplier:  Ambeed
CAS Number:  
Irritant
5,5'-Divinyl-2,2'-bipyridine 97% stabilized
Catalog # 77035-384
Supplier:  Ambeed
Supplier Number:  A638987-250MG
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle
  • Size:
    250 mg
  • Cat. No.:
    77035-384

Specifications

About this item

 :
 :208.25 g/mol
 :Stabilised with 4-tert-butylpyrocatechol (TBC)
 :GHS07
CAS Number:932396-96-8
Formula:C₁₄H₁₂N₂
MDL Number:MFCD16876215