Order Entry
Puerto Rico
ContactUsLinkComponent
3-((Dimethylamino)methyl)-5-methylhexan-2-one 97%
Irritant
3-((Dimethylamino)methyl)-5-methylhexan-2-one 97%
Catalog # 76989-794
Supplier:  Ambeed
CAS Number:  
Irritant
3-((Dimethylamino)methyl)-5-methylhexan-2-one 97%
Catalog # 76989-794
Supplier:  Ambeed
Supplier Number:  A360669-1G
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle
  • Size:
    1 g
  • Cat. No.:
    76989-794

Specifications

About this item

 :
 :171.27 g/mol
 :GHS07
CAS Number:91342-74-4
Formula:C₁₀H₂₁NO
MDL Number:MFCD22683881