Order Entry
Puerto Rico
ContactUsLinkComponent
2-(2-Ethoxy-5-((4-ethylpiperazin-1-yl)sulfonyl)phenyl)-5-methyl-7-propylimidazo[5,1-f][1,2,4]triazin-4(3H)-one 98%
Toxic
Health
Environment
2-(2-Ethoxy-5-((4-ethylpiperazin-1-yl)sulfonyl)phenyl)-5-methyl-7-propylimidazo[5,1-f][1,2,4]triazin-4(3H)-one 98%
Catalog # 77014-536
Supplier:  Ambeed
CAS Number:  
Toxic
Health
Environment
2-(2-Ethoxy-5-((4-ethylpiperazin-1-yl)sulfonyl)phenyl)-5-methyl-7-propylimidazo[5,1-f][1,2,4]triazin-4(3H)-one 98%
Catalog # 77014-536
Supplier:  Ambeed
Supplier Number:  A277692-1MG
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle
  • Size:
    1 mg
  • Cat. No.:
    77014-536

Specifications

About this item

 :
 :488.6 g/mol
UN:2811
ADR:6.1,Ⅲ
 :GHS06
 :GHS08
 :GHS09
CAS Number:224785-90-4
Formula:C₂₃H₃₂N₆O₄S
MDL Number:MFCD07773094