Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
4-Bromo-2-phenoxybenzonitrile 98%

4-Bromo-2-phenoxybenzonitrile 98%

Irritant
4-Bromo-2-phenoxybenzonitrile 98%
Supplier:  
Certificates
4-Bromo-2-phenoxybenzonitrile 98%

0 Options Available Below

  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Caution: for Research Use Only

  • CAS Number:875664-25-8
  • Formula:C₁₃H₈BrNO
  • MDL Number:MFCD13193389
  • Molecular Weight:274.11 g/mol
Product Family Options
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.