
Sodium hydroxide solution 0.5 N (0.5 M), in single-use containers, HyClone™ for bioprocessing
Supplier: Cytiva
Certificates
About this item
CAS Number:1310-73-2
:
Formula:NaOH
MDL Number:MFCD00003548
:40 g/mol
Storage Temperature:Ambient
EINECS:215-185-5
UN:1824
ADR:8,II
REACH:01-2119457892-27
:GHS05
Specification Test Results
- Storage:15° to 30°C
- Certifications/Compliance:Manufactured in a ISO9001 and ISO13485 certified facility
- Shelf life:12 Months
- pH:For Information Only
- For use with:Chromatography resins, IC systems, ÄKTA hardware systems
- Appearance:Clear, colorless solution
- Sterility:No Growth