Viral inactivation solution, Biotech Reagent, J.T.Baker®
:
:0.9 – 1.1 g/ml
:Ambient
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental Certification:EPA safer choice chemical
- ID (by FTIR):Conforms
- Appearance:Passes Test
- Purity by HPLC:min 95%
- Assay:Passes Test
- Acid value:max 10
- Viscosity (25°C):For information only
- Density:0.9 – 1.1 g/mL
- Limit of free ethylene oxide (Target <1 ppm):For information only
- Total Aerobic Microbial Count:1000 cfu/mL
- Total Yeast and Mold Count:10 cfu/mL
- Endotoxin:For information only
- Protein reactive impurities (100x Diluted):For information only
- Arsenic (As):For information only
- Cadmium (Cd):For information only
- Lead (Pb):For information only
- Mercury (Hg):For information only
- Cobalt (Co):For information only
- Nickel (Ni):For information only
- Vanadium (V):For information only
- Silver (Ag):For information only
- Gold (Au):For information only
- Iridium (Ir):For information only
- Osmium (Os):For information only
- Palladium (Pd):For information only
- Platinum (Pt):For information only
- Rhodium (Rh):For information only
- Ruthenium (Ru):For information only
- Selenium (Se):For information only
- Thallium (Tl):For information only
- Barium (Ba):For information only
- Chromium (Cr):For information only
- Copper (Cu):For information only
- Lithium (Li):For information only
- Molybdenum (Mo):For information only
- Antimony (Sb):For information only
- Tin (Sn):For information only
Avantor JTBaker Viral Inactivation Solution Poster 2025.pdf Viral Inactivation Solution, Biotech Reagent