Order Entry
Puerto Rico
ContactUsLinkComponent
Human Anti-Cytomegalovirus gB Protein Antibody IgG Titer Serologic Assay Kit
Human Anti-Cytomegalovirus gB Protein Antibody IgG Titer Serologic Assay Kit
Catalog # 77007-404
Supplier:  ReVacc Scientific
CAS Number:  
undefined
Human Anti-Cytomegalovirus gB Protein Antibody IgG Titer Serologic Assay Kit
Catalog # 77007-404
Supplier:  ReVacc Scientific
Supplier Number:  W19V42
CAS Number:  

Specifications

  • Description:
    Human anti-cytomegalovirus GB kit
  • Cat. No.:
    77007-404

Specifications

About this item

This kit is developed for serologic test for human IgG titer of Anti-HCMV gB antibody in serum/plasma or purified human antibody (monoclonal and polyclonal) in vitro.

  • This kit was tested to bind to a monoclonal antibody or convalescent human sera

Assay principles: This kit is developed for a standard indirect-ELISA format, providing a rapid detection of anti-HCMV (Towne strain) human IgG in serum. The kit consists of high-bind detachable 96-well plate pre-coated with protein, positive control, negative control, an HRP-Anti-Human IgG secondary antibody, TMB and dilution/wash/stop buffer.

Use protocol (4 simple steps):
1. Detach wells from plate based on your samples and keep at room temperature. Dilute your samples, positive and negative controls in sample dilution buffer and add them to wells (0.1 ml/well). Incubate at 37 °C for 1 hour or room temperature for 1.5 hour. Wash wells 4 times by wash solution (0.3 ml).
2. Dilute the secondary antibody HRP-Anti-Human IgG by wash solution and add to each well (0.1 ml). Incubate at room temperature for 0.5 to 1 hour. Avoid light by foil. Wash wells as mentioned.
3. Add TMB to each well (0.1 ml) and incubate at room temperature for 5 to 30 minutes.
4. Stop the substrate reaction by adding stop solution (0.1 ml/well). Absorbance (OD) is calculated as the absorbance at 450 nm. The OD value reflects the amount of antibody bound.

Storage and shipping condition: Shipment with room temperature. The unopened kit is stable for 6 months from the date of manufacture when stored at 2 to 8 °C. Avoid light. The shelf life is 60 days from the date of opening.

Caution: It is for research use only.