ContactUsLinkComponent
PTS Collect™ Capillary Tubes 40 μl
Catalog # 76994-112
Supplier:  PTS Diagnostics
undefined
PTS Collect™ Capillary Tubes 40 μl
Catalog # 76994-112
Supplier:  PTS Diagnostics
Supplier Number:  2866
Specifications
  • Description:
    PTS Collect™ capillary tubes, 40 μl
  • Cat. No.:
    76994-112
Specifications
About this item
PTS Collect™ capillary tubes provide speed, efficiency and savings especially for applications where exact sample volumes are required.
  • Ease-of-use with no assembly
  • One-hand blood collection and dosing
  • Glass tube for easy blood collection
  • Bulb-squeeze dispensing technology that ensures consistent performance
  • Only one item to inventory (no plunger)
PTS Collect capillary tubes provide better efficiency and are designed to be easier to use than traditional blood transfer tubes. Using a single glass tube with plastic casing, PTS Collect capillary tubes provide consistent self-wicking technology to preset volumes. This improved design is perfect for blood collection needs covering a wide range of applications across many different medical devices.