Order Entry
Puerto Rico
ContactUsLinkComponent
Geranyl acetate 100 µg/mL in Acetonitrile, Dr. Ehrenstorfer, LGC Standards
Geranyl acetate 100 µg/mL in Acetonitrile, Dr. Ehrenstorfer, LGC Standards
  77696-127
New Product
 :  
undefined
Geranyl acetate 100 µg/mL in Acetonitrile, Dr. Ehrenstorfer, LGC Standards
  77696-127
 :  DRE-A14010500AL-100
New Product
 :  

 

  • CAS No.:
    105-87-3
  • Description:
    Geranyl acetate 100 µg/mL in Acetonitrile
  • Size:
    1 ml
  • Formula:
    C₁₂H₂₀O₂
  • Molecular Weight:
    196.29 g/mol
  • Cat. No.:
    77696-127