Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
TerpiMix 3 2500 µg/mL in Isopropanol, Dr. Ehrenstorfer, LGC Standards
TerpiMix 3 2500 µg/mL in Isopropanol, Dr. Ehrenstorfer, LGC Standards
TerpiMix 3 2500 µg/mL in Isopropanol, Dr. Ehrenstorfer, LGC Standards
Catalog # 77677-383
Supplier:
LGC Standards
New Product
CAS Number:
Expand image
TerpiMix 3 2500 µg/mL in Isopropanol, Dr. Ehrenstorfer, LGC Standards
Catalog # 77677-383
Supplier:
LGC Standards
Supplier Number:
DRE-A50000654IP
New Product
CAS Number:
Specifications
Description:
TerpiMix 3 2500 µg/mL in Isopropanol
Size:
1 ml
Cat. No.:
77677-383
Specifications
About this item
Product Details
Product Details