Borneol (20% Isoborneol) 1000 µg/mL in Methanol, Dr. Ehrenstorfer, LGC Standards
Catalog # 77691-747
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:507-70-0
- Description:Borneol (20% Isoborneol) 1000 µg/mL in Methanol
- Size:1 ml
- Formula:C₁₀H₁₈O
- Molecular Weight:154.25 g/mol
- Cat. No.:77691-747