Limonene 1000 µg/mL in Isopropanol, Dr. Ehrenstorfer, LGC Standards
Catalog # 77696-399
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:138-86-3
- Description:Limonene 1000 µg/mL in Isopropanol
- Size:1 ml
- Formula:C₁₀H₁₆
- Molecular Weight:136.24 g/mol
- Cat. No.:77696-399