Geraniol 1000 µg/mL in Isopropanol, Dr. Ehrenstorfer, LGC Standards
Catalog # 77696-401
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:106-24-1
- Description:Geraniol 1000 µg/mL in Isopropanol
- Size:1 ml
- Formula:C₁₀H₁₈O
- Molecular Weight:154.25 g/mol
- Cat. No.:77696-401