Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
(R)-(+)-Pulegone 1000 µg/mL in Hexane, Dr. Ehrenstorfer, LGC Standards
(R)-(+)-Pulegone 1000 µg/mL in Hexane, Dr. Ehrenstorfer, LGC Standards
(R)-(+)-Pulegone 1000 µg/mL in Hexane, Dr. Ehrenstorfer, LGC Standards
77691-774
:
LGC Standards
New Product
:
(R)-(+)-Pulegone 1000 µg/mL in Hexane, Dr. Ehrenstorfer, LGC Standards
77691-774
:
LGC Standards
:
DRE-GA09011031HE
New Product
:
CAS No.:
89-82-7
Description:
(R)-(+)-Pulegone 1000 µg/mL in Hexane
Size:
1 ml
Formula:
C₁₀H₁₆O
Molecular Weight:
152.23 g/mol
Cat. No.:
77691-774