ContactUsLinkComponent
BD MAX™ StaphSR Kit
BD MAX™ StaphSR Kit
Catalog # 76906-222
Supplier:  BD
CAS Number:  
undefined
BD MAX™ StaphSR Kit
Catalog # 76906-222
Supplier:  BD
Supplier Number:  443419
CAS Number:  
Specifications
  • Description:
    BD MAX™ StaphSR test kit
  • Cat. No.:
    76906-222
  • No. of tests:
    24
Specifications
About this item
BD MAX™ StaphSR assay provides accurate and rapid detection of both staphylococcus aureus and MRSA nasal colonization in presurgical patients.
  • Staphylococcus aureus
  • MRSA
  • Molecular
Packaging: BD MAX™ vaginal pane.