ContactUsLinkComponent
BD MAX™ MRSA XT Kit
BD MAX™ MRSA XT Kit
Catalog # 76906-214
Supplier:  BD
CAS Number:  
undefined
BD MAX™ MRSA XT Kit
Catalog # 76906-214
Supplier:  BD
Supplier Number:  443461
CAS Number:  
Specifications
  • Description:
    BD MAX™ MRSA XT test kit
  • Cat. No.:
    76906-214
  • No. of tests:
    24
Specifications
About this item
The BD MAX™ MRSA XT test is a qualitative in vitro diagnostic test for detection of methicillin-resistant Staphylococcus aureus (MRSA) DNA from nasal swabs in patients at risk for nasal colonization.
  • Methicillin-resistant Staphylococcus aureus (MRSA)
  • Molecular
The test utilizes real-time polymerase chain reaction (PCR) for the amplification of MRSA DNA and fluorogenic target-specific hybridization probes for the detection of amplified DNA: specific amplicon in the SCCmec right-extremity junction (MREJ), the genes for methicillin resistance mecA and mecC, the gene nuc encoding a thermostable nuclease of S. aureus. The assay is intended to aid in the prevention and control of MRSA infections in healthcare settings.