Catalog # MSPP-SPO2-MSE
Supplier: KENT SCIENTIFIC CORP MS
Specifications
- Description:Mouse paw pulse oximeter sensors
- Environmentally Preferable:
- Cat. No.:MSPP-SPO2-MSE
Specifications
About this item
Mouse paw pulse oximeter sensors, for use with the PhysioSuite® and MouseSTAT Jr.® systems.
- CT compatible
- Designed for use on anesthetized animals