Disposable Inoculating Loops and Needles
About this item
High impact polystyrene loops and needles are smooth and flexible to facilitate uniform and smooth streaking without damaging the gel surface. Needles are straight and suitable for removal of specimens of single colonies. Packed sterile in safe, tamper proof, zip-seal resealable bags.
Disposable inoculating loops and needles do not need flaming and thus eliminate the risk of infection due to aerosol formation of pathogenic substances. Cross-contamination due to improper sterilization is eliminated. They can be used under hoods without danger, and are color-coded for ease of size identification.
2 Options Available Below
Product Details & Documents
High impact polystyrene loops and needles are smooth and flexible to facilitate uniform and smooth streaking without damaging the gel surface. Needles are straight and suitable for removal of specimens of single colonies. Packed sterile in safe, tamper proof, zip-seal resealable bags.
Disposable inoculating loops and needles do not need flaming and thus eliminate the risk of infection due to aerosol formation of pathogenic substances. Cross-contamination due to improper sterilization is eliminated. They can be used under hoods without danger, and are color-coded for ease of size identification.
Product Family Options
Product Information
- VolumeLengthColorSterilityPackagingAvailabilityPrice
- 101101-376Inoculating loopSupplier Number: 1213M74Specifications:MarketsourceMarketsource
- 101101-322Inoculating loopSupplier Number: 1213M77Specifications:MarketsourceMarketsource