Orders LinkContactUsLinkComponent
Avantor is setting science in motion for a better world | Avantor
Supplier:   Chemglass
Supplier:  Chemglass
Certificates
undefined
6 Options Available Below
Product Details & Documents
Wooden inoculation picks provide a faster way to inoculate. It’s simple, after autoclaving, just "pick" the colony, drop the wooden pick into your media and close the lid. No need to remove the pick.
  • Go from autoclave to microtube
  • Non-sterile
  • 100% USA grown White Birch
Certifications: ISO and FDA registered.
Ordering information: Available in three lengths and with semi or needle points
Product Family Options
Product Information
  • Ø
    Length
    Tip style
    Packaging
    Availability
    Price
  • 10034-204
    Inoculation pick, mini, fits 1.5 ml tubes
    Inoculation pick, mini, fits 1.5 ml tubes
    0.254 cm (ext. Ø)
    32.5 mm (1.28")
    Semi point
    Log in for availability
    Log in for availability
    Case of 2,000
    Case of 2,000 - $162.00
    $162.00
    Case of 2,000
       $162.00
    Supplier Number: CLS-3670-100
  • 10034-206
    Inoculation pick, mini, fits 1.5 ml tubes
    Inoculation pick, mini, fits 1.5 ml tubes
    0.254 cm (ext. Ø)
    32.5 mm (1.28")
    Needle point
    Log in for availability
    Log in for availability
    Case of 2,000
    Case of 2,000 - $162.00
    $162.00
    Case of 2,000
       $162.00
    Supplier Number: CLS-3670-102
  • 10034-208
    Inoculation pick, standard
    Inoculation pick, standard
    0.254 cm (ext. Ø)
    63.5 mm (2.5")
    Semi point
    1600/pack
    Log in for availability
    Log in for availability
    Pack of 1,600
    Pack of 1,600 - $225.00
    $225.00
    Pack of 1,600
       $225.00
    Supplier Number: CLS-3670-104
  • 10034-210
    Inoculation pick, standard
    Inoculation pick, standard
    0.254 cm (ext. Ø)
    63.5 mm (2.5")
    Needle point
    1600/pack
    Log in for availability
    Log in for availability
    Pack of 1,600
    Pack of 1,600 - $225.00
    $225.00
    Pack of 1,600
       $225.00
    Supplier Number: CLS-3670-106
  • 10034-212
    Inoculation pick, large
    Inoculation pick, large
    0.3175 cm (ext. Ø)
    152.4 mm (6")
    Semi point
    Log in for availability
    Log in for availability
    Pack of 500
    Pack of 500 - $127.00
    $127.00
    Pack of 500
       $127.00
    Supplier Number: CLS-3670-108
  • 10034-214
    Inoculation pick, large
    Inoculation pick, large
    0.3175 cm (ext. Ø)
    152.4 mm (6")
    Needle point
    Log in for availability
    Log in for availability
    Pack of 500
    Pack of 500 - $123.00
    $123.00
    Pack of 500
       $123.00
    Supplier Number: CLS-3670-110