Sampling Kit for Identifier System
Product Details & Documents
The sampling kit allows for the collection of respirable particles for inspection in the STAT PEEL® Identifier system.
- 6 Battery powered badges for respirable particle collection
- Tablet computer for badge control
- Badge chargers
- All necessary tools to perform air samplings
- Starter kit of 20 filtration slides
The sampling kit allows for the collection of respirable particles for inspection in the STAT PEEL® Identifier System or for SEM inspection. The included filtration badges are personal sampling devices equipped with a silent pump, environmental sensors, and filtration slides that collect respirable particles. It can be worn in a lab coat pocket to sample personal exposure or set up in a desired location to sample environmental contamination. The badge is operated with an easy-to-use app on a tablet computer. Up to 10 sampling badges can be controlled by one tablet. After completion of the sampling run, the badge data can be transferred to the tablet computer.
: CE marked.
Specifications
- Description:Sampling kit for identifier system
- Cat. No.:76848-022
- Dimensions:500×420×175 mm (19.7×16.5×6.9")
- Power:100/240 V
- Weight:6.8 kg (15 lbs.)
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



