Order Entry
Puerto Rico
ContactUsLinkComponent
Sodium chloride 0.9% USP for injection
Sodium chloride 0.9% USP for injection
Catalog # 68000-320
Supplier:  Baxter
CAS Number:  
Sodium chloride 0.9% USP for injection
Catalog # 68000-320
Supplier:  Baxter
Supplier Number:  2B1306
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Bag, Viaflex®, single pack
  • Size:
    50 mL
  • Cat. No.:
    68000-320
  • Supplier no.:
    2B1306

Specifications

About this item

 :
 :58.44 g/mol
CAS Number:7647-14-5
Formula:NaCl
MDL Number:MFCD00003477