Ringer lactate solution USP for injection
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Lactated Ringer's Injection USP is sterile, nonpyrogenic and contains no bacteriostatic or antimicrobial agents. This product is intended for intravenous administration in a single dose container.
- NDC number: 00264-7750-20
This solution is indicated for use in adults and pediatric patients as a source of electrolytes and water for hydration.
Not made with latex, PVC or DEHP.
Specific Gravity: 1.006
Specifications
Specifications
- Pack type:Container, Excel® IV
- Size:250 ml