Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Cells
MULTISCREEN™ Human P2Y2 HEK293T HTS Membranes
MULTISCREEN™ Human P2Y2 HEK293T HTS Membranes
:
MULTISPAN INC
MULTISCREEN™ Human P2Y2 HEK293T HTS Membranes
:
MULTISPAN INC
Storage Conditions:
LN₂ (Below –135 ℃)
MULTISCREEN™ Membranes Expressing GPCRs and Beyond
Multispan MULTISCREEN™ Products and Services