$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Cells
MULTISCREEN™ Human GHRH HEK293T HTS Membranes
MULTISCREEN™ Human GHRH HEK293T HTS Membranes
:
MULTISPAN INC
MULTISCREEN™ Human GHRH HEK293T HTS Membranes
:
MULTISPAN INC
Storage Conditions:
LN₂ (Below –135 ℃)
MULTISCREEN™ Membranes Expressing GPCRs and Beyond
Multispan MULTISCREEN™ Products and Services