About this item
The UVP202001 and UVP202002, Ultraviolet Laser Microscope Slide and Tissue Cassette Printer, represent the next generation of histology printers. These advanced and easy to use printers can print directly on slides cassettes without using ink or consumables.
- Next generation ultraviolet laser print technology for quick and consumable-free printing
- Large 10-inch color touch screen for data entry and print configuration
- Quick and quiet print capabilities with slides being printed in under three seconds
- Removable feed and dispensing slide trays for optimizing lab workflow
- Built-in scanner for quick reading of barcodes and data entry
The ultra-fast laser can print text, symbols, and barcodes on cassettes in under three seconds. The large 10-inch color touch screen allows for configuration of the print template and easy data entry. All Four E’s USA’s printers are LIMS and HIMS compatible for easy integration.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 76924-036
- 76924-038
- Compatibility:
- LIMS/HIMS
- LIMS/HIMS
- Display:
- 10" color touch screen
- 10" color touch screen