ContactUsLinkComponent
AccuForge® GEM® Single Edge Blade Cartridge, Coated
AccuForge® GEM® Single Edge Blade Cartridge, Coated
Catalog # 76717-308
Supplier:  Accutec Blades
undefined
AccuForge® GEM® Single Edge Blade Cartridge, Coated
Catalog # 76717-308
Supplier:  Accutec Blades
Supplier Number:  AGBL-7015-0000
Specifications
  • Coating:
    Coated
  • Description:
    Oiled
  • Material:
    Stainless steel
  • Thickness:
    0.009"
  • Packed:
    100 blades/carton, 10 cartons/case
  • Cat. No.:
    76717-308
  • Blade Edge:
    3 facet
  • Backing material:
    Steel
Specifications
About this item
AccuForge single edge blades feature 0.009 stainless steel and 3-facet edges for sharpness. The MicroCoat® Enrichment Technology reduces cut force and increases durability.
  • Blade edge with MicroCoat® technology enhances durability and sharpness
  • Cartridge inserts into reloadable safety dispenser
  • Individually packed in clamshell with product key and instructions for use
  • Paper-free packaging is clean room ready
  • Each cartridge contains 100 blades
AccuForge's MicroCoat® blade enrichment technology adds a layer of protection for each blade helping maintain a sharp effective edge over multiple uses.
Ordering information: For use with GEM RUNNER™ Dispenser Cat no. 76717-342.
Caution: Blades are extremely sharp. Wear eye protection. Handle and use with care. Bending blades may break them. Keep out of children’s reach. Dispose of safely.