ContactUsLinkComponent
EVOLUTION* 4 Surgical Sleeve, Sterile
EVOLUTION* 4 Surgical Sleeve, Sterile
  76836-638
undefined
EVOLUTION* 4 Surgical Sleeve, Sterile
  76836-638
 :  89791
  • Ø:
    15 cm (6")
  • Sterile:
    +
  • Color:
    Blue
  • Description:
    EVOLUTION* 4 surgical sleeves
  • Length:
    59 cm (23.5")
  • Cat. No.:
    76836-638
EVOLUTION* 4 surgical sleeves are available in a universal/one-size fit.
  • Low linting
  • Flame-resistant
  • Abrasion-resistant
Blue SMS fabric surgical sleeves are supplied sterile.
 : Meets NFPA Class I and ISO 11810. Meets ASTM D4966.