Order Entry
United States
ContactUsLinkComponent
Mag-Bind® Total RNA 96 Kit
Mag-Bind® Total RNA 96 Kit
Catalog # 76321-016
Supplier:  Omega Bio-Tek
CAS Number:  
Mag-Bind® Total RNA 96 Kit
Catalog # 76321-016
Supplier:  Omega Bio-Tek
Supplier Number:  M6731-00
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Application:
  • Method / Format:
    Magnetic beads
  • Prep size:
    Scalable
  • Refine target molecule:
    Total RNA
  • Sample type:
    Cultured cells
    Human/animal cells and tissues
  • Target molecule:
    RNA
  • Description:
    Total RNA kit
  • Size:
    1×96 preps
  • Environmentally Preferable:
  • Cat. No.:
    76321-016

Specifications

About this item

Isolate total cellular RNA and viral RNA from a wide variety of cells and tissues.

  • Procedure can be fully automated with most robotic workstations

The Mag-Bind® Total RNA 96 Kit provides a novel technology for total RNA isolation. This kit allows the rapid and reliable isolation of high-quality total cellular RNA and viral RNA from a wide variety of cells and tissues. Unlike column-based systems, the binding of nucleic acids to magnetic particles occurs in solution resulting in increased binding kinetics and binding efficiency. Particles are also completely re-suspended during the wash steps of the purification protocol, which improves the removal of contaminants and increases nucleic acid purity. Mag-Bind® Total RNA 96 Kit procedure can be fully automated with most robotic workstations.