SATWipes® Replacement Canister
SATWipes® replacement canister, 15.2×22.9 cm (6×9")
Supplier: Contec
Supplier #: CSCN0100
Jump to:
Product Details & Documents
A replacement canister for the SATWipes® 6×9" rolls saturated with IPA greater than 70%.
- Canister features a reinforced snap-close lid
Documents
Certificates
Search by Lot NumberSpecifications
- Description:SATWipes® replacement canister
- W×L:15.2×22.9 cm (6×9")
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



