Order Entry
Puerto Rico
ContactUsLinkComponent
Ethyl pyruvate 98%
Flammable
Ethyl pyruvate 98%
Catalog # 76844-868
Supplier:  Ambeed
CAS Number:  
Flammable
Ethyl pyruvate 98%
Catalog # 76844-868
Supplier:  Ambeed
Supplier Number:  A703267-25G
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle
  • Size:
    25 g
  • Cat. No.:
    76844-868

Specifications

About this item

 :
 :116.12 g/mol
UN:3272
ADR:3,Ⅲ
 :GHS02
CAS Number:617-35-6
Formula:C₅H₈O₃
MDL Number:MFCD00009123