Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-Infliximab Mouse Monoclonal Antibody [clone: 23H5]
Anti-Infliximab Mouse Monoclonal Antibody [clone: 23H5]
Catalog # 76510-102
Supplier:  Genscript
Anti-Infliximab Mouse Monoclonal Antibody [clone: 23H5]
Catalog # 76510-102
Supplier:  Genscript
Supplier Number:  A02041-40

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Infliximab
  • Antigen Symbol:
    Infliximab
  • Clonality:
    Monoclonal
  • Clone:
    23H5
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Size:
    40 µg
  • Cross Adsorption:
    No
  • Form:
    Lyophilized
  • Storage Buffer:
    PBS, pH 7.4, containing 0.02% sodium azide
  • Storage Temperature:
    The lyophilized product remains stable up to 1 year at −20 °C from date of receipt. Upon reconstitution, it can be stored for 2 - 3 weeks at 2 - 8 °C or for up to 12 months at −20 °C or below. Avoid repeated freezing and thawing cycles
  • Concentration:
    Reconstitute the lyophilized powder with deionized water (or equivalent) to a final concentration of 0.5 mg/ml
  • Shipping Temperature:
    Ice
  • Immunogen:
    Infliximab
  • Purification:
    Protein A affinity column
  • Cat. No.:
    76510-102
  • Packaging:
    Vial

Specifications

About this item

The product is specific for infliximab. The antibody is recommended as a detection antibody in a pharmacokinetic (PK) bridging assay with capture antibody GenScript, A02040-40, anti-infliximab antibody (27A4), mAb, mouse.

  • Anti idiotype antibodies
  • High specificit
  • High affinity
  • High stability

GenScript has developed a broad range of off-the-shelf anti-idiotype antibodies against blockbuster drugs to help accelerate your research of biosimilars.
.

Type: Primary
Antigen: Infliximab
Clonality: Monoclonal
Clone: 23H5
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: