Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-SARS-CoV-2 Spike S1 Human Monoclonal Antibody [clone: hIgA2001]
Anti-SARS-CoV-2 Spike S1 Human Monoclonal Antibody [clone: hIgA2001]
Supplier:  Genscript
Certificates
Anti-SARS-CoV-2 Spike S1 Human Monoclonal Antibody [clone: hIgA2001]
Supplier:  Genscript

Specifications

  • Catalog No:
  • 76510-174
  • 76510-172
  • Antigen Symbol:
  • SARS-CoV-2 Spike S1
  • SARS-CoV-2 Spike S1
  • Antigen Name:
  • SARS-CoV-2 Spike S1
  • SARS-CoV-2 Spike S1
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • hIgA2001
  • hIgA2001
  • Host:
  • Human
  • Human
  • Isotype:
  • IgA
  • IgA
  • ELISA:
  • Yes
  • Yes
  • Immunogen:
  • Recombinant SARS-CoV-2 RBD Protein
  • Recombinant SARS-CoV-2 RBD Protein
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • Affinity chromatography
  • Affinity chromatography
  • Concentration:
  • 1 mg/ml
  • 1 mg/ml
  • Cross Adsorption:
  • No
  • No
  • Storage Buffer:
  • PBS, pH 7.4, containing 0.02% sodium azide
  • PBS, pH 7.4, containing 0.02% sodium azide
  • Storage Temperature:
  • Store at −20 °C. This product is stable for 1 year upon receipt, when handled and stored as instructed. Avoid repeated freezing and thawing cycles
  • Store at −20 °C. This product is stable for 1 year upon receipt, when handled and stored as instructed. Avoid repeated freezing and thawing cycles
  • Shipping Temperature:
  • Ice
  • Ice

Specifications