Anti-SARS-CoV-2 Spike S1 Human Monoclonal Antibody [clone: hIgM2001]
Supplier: Genscript
Certificates
About this item
The product is specific for SARS-CoV-2 spike protein S1 subunit and its RBD domain.
- COVID-19 antibodies
- Validated for ELISA, WB, FACS, and SARS-CoV-2 virus neutralization assay
- Control antibodies and various antibody pairs available for ELISA based detection
- Neutralizing antibody standard to calibrate quantitative assay of SARS-CoV-2 neutralizing antibodies in samples
Genscript offers site-specific neutralizing antibodies, neutralizing antibody standards, and control antibodies.
Type: Primary
Antigen: SARS-CoV-2 Spike S1
Clonality: Monoclonal
Clone: hIgM2001
Conjugation:
Epitope:
Host: Human
Isotype: IgM
Reactivity:
Specifications
- Catalog No:
- 76510-110
- 76510-112
- Antigen Symbol:
- SARS-CoV-2 Spike S1
- SARS-CoV-2 Spike S1
- Antigen Name:
- SARS-CoV-2 Spike S1
- SARS-CoV-2 Spike S1
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- hIgM2001
- hIgM2001
- Host:
- Human
- Human
- Isotype:
- IgM
- IgM
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant SARS-CoV-2 RBD Protein
- Recombinant SARS-CoV-2 RBD Protein
- Form:
- Liquid
- Liquid
- Purification:
- Affinity chromatography
- Affinity chromatography
- Concentration:
- 1 mg/ml
- 1 mg/ml
- Cross Adsorption:
- No
- No
- Storage Buffer:
- PBS, pH 7.4, containing 0.02% sodium azide
- PBS, pH 7.4, containing 0.02% sodium azide
- Storage Temperature:
- Store at −20 °C. This product is stable for 1 year upon receipt, when handled and stored as instructed. Avoid repeated freezing and thawing cycles
- Store at −20 °C. This product is stable for 1 year upon receipt, when handled and stored as instructed. Avoid repeated freezing and thawing cycles
- Shipping Temperature:
- Ice
- Ice