Anti-Rituximab Rabbit Monoclonal Antibody [clone: 137C6]
Catalog # 76509-912
Supplier: Genscript
Specifications
- Antibody Type:Primary
- Antigen Name:Rituximab
- Antigen Symbol:Rituximab
- Clonality:Monoclonal
- Clone:137C6
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Size:40 µg
- Cross Adsorption:No
- Form:Lyophilized
- Storage Buffer:PBS, pH 7.4, contains 0.02% sodium azide
- Storage Temperature:The lyophilized product remains stable up to 1 year at −20 °C from date of receipt. Upon reconstitution, it can be stored for 2 - 3 weeks at 2 - 8 °C or for up to 12 months at −20 °C or below. Avoid freeze/thaw cycles
- Concentration:Reconstitute the lyophilized powder with deionized water (or equivalent) to a final concentration of 0.5 mg/ml
- Shipping Temperature:Ice
- Immunogen:Rituximab
- Purification:Protein A affinity column
- Cat. No.:76509-912
- Packaging:Vial
Specifications
About this item
The product is specific for Rituximab. The antibody is recommended as a capture antibody in a pharmacokinetic (PK) bridging assay with detection antibody GenScript, A01944-40, MonoRab™ Anti-Rituximab Antibody (194D7) [Biotin], mAb, Rabbit.
- Anti-Idiotype antibodies
- High specificit
- High affinity
- High stability
GenScript has developed a broad range of off-the-shelf anti-idiotype antibodies against blockbuster drugs to help accelerate your research of biosimilars.
Type: Primary
Antigen: Rituximab
Clonality: Monoclonal
Clone: 137C6
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: