Order Entry
Puerto Rico
ContactUsLinkComponent
 
Anti-Camelid VHH Rabbit Antibody
  76510-044
 :  Genscript
Anti-Camelid VHH Rabbit Antibody
  76510-044
 :  Genscript
 :  A02014-200
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Antibody Type:
    Primary
  • Antigen Name:
    Camelid VHH
  • Antigen Symbol:
    Camelid VHH
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Western Blot:
    Yes
  • Size:
    200 µl
  • Cross Adsorption:
    No
  • Form:
    Liquid
  • Storage Buffer:
    PBS (pH 7.4) with 0.02% sodium azide and 50% glycerol
  • Storage Temperature:
    Store at −20 °C. This product is stable for 1 year upon receipt, when handled and stored as instructed
  • Concentration:
    0.5 mg/ml
  • Shipping Temperature:
    Ice
  • Immunogen:
    Llama VHH antibody
  • Purification:
    Protein A affinity column
  • Cat. No.:
    76510-044
  • Packaging:
    Vial

 

 

GenScript’s MonoRab™ Rabbit Anti-Camelid VHH Cocktail is specific for Camelid VHH antibody (Llama, Alpaca, and Camel). It has no cross-reactivity with mouse, rat, rabbit, goat or human immunoglobulins.

  • Anti-VHH Antibodies
  • MonoRab™ technology guarantees high affinity: Kd ~10 to 11 M
  • No cross reactivity with other species
  • Specific to camelid IgG2 and IgG3
  • Recognize conformational epitopes of the VHH domain
  • Recognize both camelid and humanized VHHs

GenScript has developed a series of rabbit monoclonal (MonoRab) anti-VHH antibodies to facilitate your research of VHHs. Our anti-VHH antibodies recognize conformational epitopes of the VHH domain of camelid lgG2 and lgG3 subclasses. It is not necessary to add a tag to VHH for detection anymore.

Type: Primary
Antigen: Camelid VHH
Clonality:
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: