Anti-Camelid VHH Rabbit Monoclonal Antibody [clone: 96A3F5] (iFluor 555)
Catalog # 76197-322
Supplier: Genscript
Specifications
- Antibody Type:Primary
- Antigen Name:Camelid VHH
- Antigen Symbol:Camelid VHH
- Clonality:Monoclonal
- Clone:96A3F5
- Conjugation:iFluor® 555
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Camel,Llama,Alpaca
- Western Blot:Yes
- Size:200 µl
- Cross Adsorption:No
- Form:Liquid
- Storage Buffer:PBS (pH 7.4), 10 mg/ml BSA, 0.02% sodium azide and 50% glycerol
- Storage Temperature:Store at −20 °C. This product is stable for 1 year upon receipt, when handled and stored as instructed
- Concentration:0.5 mg/ml
- Shipping Temperature:Ice
- Immunogen:Llama VHH antibody
- Purification:Protein A affinity column
- Cat. No.:76197-322
- Packaging:Vial
Specifications
About this item
GenScript MonoRab™ Rabbit Anti-Camelid VHH Antibody [iFluor555], mAb is specific for Camelid VHH antibody (Alpaca, Llama and Camel). The monoclonal antibody has no cross-reactivity with mouse, rat, rabbit, chicken, goat or human immunoglobulins.
- Anti-VHH Antibodies
- MonoRab™ technology guarantees high affinity: Kd ~10 to 11 M
- No cross reactivity with other species
- Specific to camelid IgG2 and IgG3
- Recognize conformational epitopes of the VHH domain
- Recognize both camelid and humanized VHHs
GenScript has developed a series of rabbit monoclonal (MonoRab) anti-VHH antibodies to facilitate your research of VHHs. Our anti-VHH antibodies recognize conformational epitopes of the VHH domain of camelid lgG2 and lgG3 subclasses. It is not necessary to add a tag to VHH for detection anymore.
Type: Primary
Antigen: Camelid VHH
Clonality: Monoclonal
Clone: 96A3F5
Conjugation: iFluor 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Alpaca;Llama;Camel