Specifications
- Antibody Type:Secondary
- Antigen Name:IgG Fc
- Antigen Symbol:IgG Fc
- Clonality:Monoclonal
- Clone:14H9H10
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Rabbit
- Western Blot:Yes
- Size:200 µl
- Cross Adsorption:No
- Form:Liquid
- Storage Buffer:PBS (pH 7.4), 10 mg/ml BSA, 0.01% thimerosal and 50% glycerol
- Storage Temperature:Store at −20 °C. This product is stable for 1 year upon receipt, when handled and stored as instructed
- Shipping Temperature:Ice
- Immunogen:Rabbit IgG(H and L)
- Purification:Protein A affinity column
- Cat. No.:76197-308
- Packaging:Vial
Specifications
About this item
GenScript Mouse Anti-Rabbit IgG Fc Antibody (14H9H10)[HRP], mAb reacts with the Fc portion of rabbit IgG but not with the Fab portion of rabbit IgG. The monoclonal antibody has no cross-reactivity with human, mouse, rat, sheep, bovine, goat or camel immunoglobulins.
- High specificit
- High affinity
- High stability
At GenScript, secondary antibodies are purified from the serum of several hosts, including goat, donkey, mouse, rabbit, chicken and sheep. Secondary antibodies are available with specificity for whole Ig molecules or antibody fragments such as the Fc or Fab regions. The conjugated secondary antibodies can be used for various immunology analysis and protein assays.
Type: Secondary
Antigen: IgG Fc
Clonality: Monoclonal
Clone: 14H9H10
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Rabbit