Order Entry
Puerto Rico
ContactUsLinkComponent
 
Irrigation Set, TUR/Bladder Y-Type, Regulating Clamp, Baxter
  68000-730
 :  Baxter
Irrigation Set, TUR/Bladder Y-Type, Regulating Clamp, Baxter
  68000-730
 :  Baxter
 :  2C4041

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • Description:
    Y-Type TUR/Bladder Irrigation Set, length 82" (2.1 m), Sterile, Non-pyrogenic
  • Cat. No.:
    68000-730
  • Supplier no.:
    2C4041

 

 

For use with Baxter irrigating solutions in plastic containers such as UROMATIC or ARTHROMATIC plastic containers.

  • Contains DEHP
  • Sterile Package
  • Non pyrogenic

Approximate Length 82" (2.1 m)