Order Entry
Puerto Rico
ContactUsLinkComponent
Chicken Luteinizing Hormone Releasing Hormone ELISA Kit
Chicken Luteinizing Hormone Releasing Hormone ELISA Kit
Catalog # 76745-886
Supplier:  ANTIBODIES.COM LLC
CAS Number:  
undefined
Chicken Luteinizing Hormone Releasing Hormone ELISA Kit
Catalog # 76745-886
Supplier:  ANTIBODIES.COM LLC
Supplier Number:  A73849-96
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Competitive
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Chicken
  • Target protein:
    luteinizing hormone releasing hormone
  • Description:
    Chicken luteinizing hormone releasing hormone ELISA kit
  • Size:
    96 tests
  • Sample Type:
    Serum, plasma, tissue homogenates and other biological fluids
  • Cross Reactivity:
    Chicken luteinizing hormone releasing hormone ELISA Kit exhibits high specificity and excellent specificity for the detection of chicken luteinizing hormone releasing hormone. No significant cross-reactivity or interference between luteinizing hormone releasing hormone and analogues was observed.
  • Detection Method:
    Colorimetric
  • Time to Results:
    4 h 30 min
  • Assay Principle:
    Quantitative
  • Shelf Life:
    Store for 6 months at 4 °C
  • Detection Range:
    31.25 - 2000 pg/ml
  • Storage Temperature:
    4 °C
  • Sample Volume:
    50 μl
  • Sensitivity:
    18.75 pg/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    76745-886

Specifications

About this item

Chicken luteinizing hormone releasing hormone ELISA kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determination of chicken luteinizing hormone releasing hormone in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready To Use ELISA Kit
  • Detection Range: 31.25 to 2000 pg/ml
  • Sensitivity: 18.75 pg/ml
  • Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
  • Assay Precision: Intra - Assay: CV <8%, Inter - Assay: CV <10%

Chicken Luteinizing Hormone Releasing Hormone ELISA Kit (A73849) employs the competitive enzyme immunoassay technique for the quantitative measurement of chicken Luteinizing Hormone Releasing Hormone in serum, plasma, tissue homogenates, and other biological fluids. The 96-well microtiter plate has been pre-coated with Luteinizing Hormone Releasing Hormone antigen. During the incubation, Luteinizing Hormone Releasing Hormone present in the samples or standards competes with the fixed amount of immobilized Luteinizing Hormone Releasing Hormone for binding sites on the Biotinylated Anti-Luteinizing Hormone Releasing Hormone Antibody. The more Luteinizing Hormone Releasing Hormone present in a sample or standard, the less Biotinylated Anti-Luteinizing Hormone Releasing Hormone Antibody that binds to the plate. Following incubation, unbound Biotinylated Anti-Luteinizing Hormone Releasing Hormone Antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is inversely proportional to the amount of Luteinizing Hormone Releasing Hormone present in each sample or standard. The concentration of Luteinizing Hormone Releasing Hormone can then be calculated by reading the O.D. absorbance at 450nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilized standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.