- Assay duration:Multiple steps
- Assay Type (ELISA with LOV):Sandwich
- Format:Pre-coated
- Primary antibody reactivity:Mouse
- Target protein:ZNF179/BFP
- Description:Mouse ZNF179/BFP ELISA kit
- Size:96 tests
- Sample Type:Serum, plasma, tissue homogenates and other biological fluids
- Cross Reactivity:Mouse ZNF179/BFP ELISA kit exhibits high specificity and excellent specificity for the detection of mouse ZNF179/BFP. No significant cross-reactivity or interference between ZNF179/BFP and analogues was observed.
- Detection Method:Colorimetric
- Time to Results:4 h 30 min
- Assay Principle:Quantitative
- Shelf Life:Store for 6 months at 4 °C
- Detection Range:0.156 - 10 ng/ml
- Storage Temperature:4 °C
- Sample Volume:100 μl
- Sensitivity:0.094 ng/ml
- Regulatory Status:RUO
- Cat. No.:76744-366
Mouse ZNF179/BFP ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse ZNF179/BFP in serum, plasma, tissue homogenates, and other biological fluids.
- Ready to use ELISA kit
- Detection range: 0.156 to 10 ng/ml
- Sensitivity: 0.094 ng/ml
- Sample type: Serum, plasma, tissue homogenates, and other biological fluids
- Assay precision: Intra assay: CV <8%, Inter assay: CV <10%
Mouse ZNF179/BFP ELISA kit (A77750) employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse ZNF179/BFP in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for ZNF179/BFP has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the ZNF179/BFP present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-ZNF179/BFP Antibody, which binds the captured ZNF179/BFP present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of ZNF179/BFP captured in each well. The concentration of ZNF179/BFP can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.