Order Entry
Puerto Rico
ContactUsLinkComponent
Human Histone H2B Type 2-E ELISA Kit
Human Histone H2B Type 2-E ELISA Kit
Catalog # 76743-486
Supplier:  ANTIBODIES.COM LLC
CAS Number:  
undefined
Human Histone H2B Type 2-E ELISA Kit
Catalog # 76743-486
Supplier:  ANTIBODIES.COM LLC
Supplier Number:  A74198-96
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Sandwich
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Human
  • Target protein:
    Histone H2B type 2-E
  • Description:
    Human histone H2B type 2-E ELISA kit
  • Size:
    96 tests
  • Sample Type:
    Serum, plasma, tissue homogenates and other biological fluids
  • Cross Reactivity:
    Human Histone H2B type 2-E ELISA Kit exhibits high specificity and excellent specificity for the detection of human Histone H2B type 2-E. No significant cross-reactivity or interference between Histone H2B type 2-E and analogues was observed.
  • Detection Method:
    Colorimetric
  • Time to Results:
    4 h 30 min
  • Assay Principle:
    Quantitative
  • Shelf Life:
    Store for 6 months at 4 °C
  • Detection Range:
    0.313 - 20 ng/ml
  • Storage Temperature:
    4 °C
  • Sample Volume:
    100 μl
  • Sensitivity:
    0.188 ng/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    76743-486

Specifications

About this item

Human Histone H2B type 2-E ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Histone H2B type 2-E in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready To Use ELISA Kit
  • Detection Range: 0.313 to 20 ng/ml
  • Sensitivity: 0.188 ng/ml
  • Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
  • Assay Precision: Intra - Assay: CV <8%, Inter - Assay: CV <10%

Human Histone H2B Type 2-E ELISA Kit (A74198) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human Histone H2B Type 2-E in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Histone H2B Type 2-E has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Histone H2B Type 2-E present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Histone H2B Type 2-E Antibody, which binds the captured Histone H2B Type 2-E present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Histone H2B Type 2-E captured in each well. The concentration of Histone H2B Type 2-E can then be calculated by reading the O.D. absorbance at 450nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilized standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.