Specifications
- Assay duration:Multiple steps
- Assay Type (ELISA with LOV):Sandwich
- Format:Pre-coated
- Primary antibody reactivity:Other
- Target protein:SARS-CoV-2 Spike Protein (RBD)
- Description:SARS-CoV-2 spike protein (RBD) ELISA kit
- Size:96 tests
- Sample Type:Serum, plasma, tissue homogenates, and other biological fluids
- Cross Reactivity:SARS-CoV-2 Spike Protein (RBD) ELISA Kit exhibits high specificity and excellent specificity for the detection of universal SARS-CoV-2 Spike Protein (RBD). No significant cross-reactivity or interference between SARS-CoV-2 Spike Protein (RBD) and analogues was observed.
- Detection Method:Colorimetric
- Time to Results:4 h 30 min
- Shelf Life:Store for 6 months at 4 °C
- Detection Range:23.438 - 1500 pg/ml
- Storage Temperature:4 °C
- Sample Volume:100 μl
- Sensitivity:14.063 pg/ml
- Regulatory Status:RUO
- Cat. No.:76743-026
Specifications
About this item
SARS-CoV-2 Spike Protein (RBD) ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of SARS-CoV-2 Spike Protein (RBD) in serum, plasma, tissue homogenates, and other biological fluids.
- Ready-to-use ELISA kit
- Assay precision: Intra-assay: CV <8%, Inter-assay: CV <10%
SARS-CoV-2 Spike Protein (RBD) ELISA kit (A303895) employs the sandwich enzyme immunoassay technique for the quantitative measurement of universal SARS-CoV-2 Spike Protein (RBD) in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for SARS-CoV-2 Spike Protein (RBD) has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the SARS-CoV-2 Spike Protein (RBD) present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-SARS-CoV-2 Spike Protein (RBD) Antibody, which binds the captured SARS-CoV-2 Spike Protein (RBD) present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of SARS-CoV-2 Spike Protein (RBD) captured in each well. The concentration of SARS-CoV-2 Spike Protein (RBD) can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.