- Assay duration:Multiple steps
- Assay Type (ELISA with LOV):Sandwich
- Format:Pre-coated
- Primary antibody reactivity:Rat
- Target protein:CX3CR1
- Description:Rat CX3CR1 ELISA kit
- Size:96 tests
- Sample Type:Serum, plasma, tissue homogenates and other biological fluids
- Cross Reactivity:Rat CX3CR1 ELISA Kit exhibits high specificity and excellent specificity for the detection of rat CX3CR1. No significant cross-reactivity or interference between CX3CR1 and analogues was observed.
- Detection Method:Colorimetric
- Time to Results:4 h 30 min
- Assay Principle:Quantitative
- Shelf Life:Store for 6 months at 4 °C
- Detection Range:23.438 - 1500 pg/ml
- Storage Temperature:4 °C
- Sample Volume:100 μl
- Sensitivity:14.063 pg/ml
- Regulatory Status:RUO
- Cat. No.:76742-336
Rat CX3CR1 ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat CX3CR1 in serum, plasma, tissue homogenates, and other biological fluids.
- Ready To Use ELISA kit
- Detection Range: 23.438 to 1500 pg/ml
- Sensitivity: 14.063 pg/ml
- Sample Type: Serum, plasma, tissue homogenates and other biological fluids
- Assay Precision: Intra - Assay: CV <8%, Inter - Assay: CV <10%
Rat CX3CR1 ELISA kit (A79022) employs the sandwich enzyme immunoassay technique for the quantitative measurement of rat CX3CR1 in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for CX3CR1 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the CX3CR1 present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-CX3CR1 Antibody, which binds the captured CX3CR1 present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of CX3CR1 captured in each well. The concentration of CX3CR1 can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.