Order Entry
Puerto Rico
ContactUsLinkComponent
Mouse Cdk6 ELISA Kit
Mouse Cdk6 ELISA Kit
Catalog # 76741-626
Supplier:  ANTIBODIES.COM LLC
CAS Number:  
undefined
Mouse Cdk6 ELISA Kit
Catalog # 76741-626
Supplier:  ANTIBODIES.COM LLC
Supplier Number:  A303429-96
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Sandwich
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Mouse
  • Target protein:
    Cdk6
  • Description:
    Mouse Cdk6 ELISA kit
  • Size:
    96 tests
  • Sample Type:
    Serum, plasma, tissue homogenates, and other biological fluids
  • Cross Reactivity:
    Mouse Cdk6 ELISA Kit exhibits high specificity and excellent specificity for the detection of mouse Cdk6. No significant cross-reactivity or interference between Cdk6 and analogues was observed.
  • Detection Method:
    Colorimetric
  • Time to Results:
    4 h 30 min
  • Assay Principle:
    Quantitative
  • Shelf Life:
    Store for 6 months at 4 °C
  • Detection Range:
    15.625 - 1000 pg/ml
  • Storage Temperature:
    4 °C
  • Sample Volume:
    100 μl
  • Sensitivity:
    9.375 pg/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    76741-626

Specifications

About this item

Mouse Cdk6 ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse Cdk6 in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready-to-use ELISA kit
  • Detection Range: 15.625 to 1000 pg/ml
  • Sensitivity: 9.375 pg/ml
  • Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
  • Assay Precision: Intra to Assay: CV <8%, Inter to Assay: CV <10%

Mouse Cdk6 ELISA kit (A303429) employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse Cdk6 in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Cdk6 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Cdk6 present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated anti-Cdk6 antibody, which binds the captured Cdk6 present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Cdk6 captured in each well. The concentration of Cdk6 can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, lyophilized standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.