- Assay duration:Multiple steps
- Assay Type (ELISA with LOV):Sandwich
- Format:Pre-coated
- Primary antibody reactivity:Rat
- Target protein:Lactate Dehydrogenase B/LDH-B
- Description:Rat lactate dehydrogenase B/LDH-B ELISA kit
- Size:96 tests
- Sample Type:Serum, plasma, tissue homogenates and other biological fluids
- Cross Reactivity:Rat Lactate Dehydrogenase B/LDH-B ELISA Kit exhibits high specificity and excellent specificity for the detection of rat Lactate Dehydrogenase B/LDH-B. No significant cross-reactivity or interference between Lactate Dehydrogenase B/LDH-B and analogues was observed.
- Detection Method:Colorimetric
- Time to Results:4 h 30 min
- Assay Principle:Quantitative
- Shelf Life:Store for 6 months at 4 °C
- Detection Range:1.563 - 100 ng/ml
- Storage Temperature:4 °C
- Sample Volume:100 μl
- Sensitivity:0.938 ng/ml
- Regulatory Status:RUO
- Cat. No.:76741-506
Rat Lactate Dehydrogenase B/LDH-B ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat Lactate Dehydrogenase B/LDH-B in serum, plasma, tissue homogenates, and other biological fluids.
- Ready To Use ELISA kit
- Detection Range: 1.563 to 100 ng/ml
- Sensitivity: 0.938 ng/ml
- Sample Type: Serum, plasma, tissue homogenates and other biological fluids
- Assay Precision: Intra - Assay: CV <8%, Inter - Assay: CV <10%
Rat Lactate Dehydrogenase B/LDH-B ELISA kit (A80059) employs the sandwich enzyme immunoassay technique for the quantitative measurement of rat Lactate Dehydrogenase B/LDH-B in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Lactate Dehydrogenase B/LDH-B has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Lactate Dehydrogenase B/LDH-B present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Lactate Dehydrogenase B/LDH-B Antibody, which binds the captured Lactate Dehydrogenase B/LDH-B present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Lactate Dehydrogenase B/LDH-B captured in each well. The concentration of Lactate Dehydrogenase B/LDH-B can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.